Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap3
Entrez Gene ID
312720
Gene Name
A kinase (PRKA) anchor protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 159399686 - 159425512 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005557.1 NP_001005557.1 3 - 4 2515 129
XM_006237488.2 XP_006237550.1 4 - 5 2700 129
XM_006237489.2 XP_006237551.1 5 - 6 2811 129
XM_006237491.2 XP_006237553.1 4 - 5 2957 129
XM_006237492.2 XP_006237554.1 4 - 5 2584 129
XM_008763287.1 XP_008761509.1 4 - 5 2547 129
XM_008763288.1 XP_008761510.1 4 - 5 2619 129
XM_008763289.1 XP_008761511.1 3 - 4 2518 129
XM_008763290.1 XP_008761512.1 3 - 4 2497 129
GenBank mRNA BC081924.1 - 3 - 4 2515 129

View Full Product Details