Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
303353
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
-
Chromosome Location
Chr.10: 67810655 - 67857562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107027.1 NP_001100497.1 11 - 12 2000 125

View Full Product Details