Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
303353
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
-
Chromosome Location
Chr.10: 67810655 - 67857562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107027.1 NP_001100497.1 2 - 3 639 67
XM_006246987.2 XP_006247049.1 2 - 3 629 67
XM_006246988.2 XP_006247050.1 2 - 3 629 67
GenBank mRNA BC168749.1 - 1 - 2 108 67
FQ225888.1 - 1 - 2 130 67

View Full Product Details