Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
303353
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
-
Chromosome Location
Chr.10: 67810655 - 67857562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107027.1 NP_001100497.1 5 - 6 936 145
XM_006246987.2 XP_006247049.1 5 - 6 926 145
XM_006246988.2 XP_006247050.1 5 - 6 926 145
GenBank mRNA BC168749.1 - 4 - 5 405 145
FQ225888.1 - 4 - 5 427 145

View Full Product Details