Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
303353
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
-
Chromosome Location
Chr.10: 67810655 - 67857562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246987.2 XP_006247049.1 12 - 13 1613 119
XM_006246988.2 XP_006247050.1 12 - 13 1613 119
GenBank mRNA BC168749.1 - 11 - 12 1092 119
FQ225888.1 - 11 - 12 1116 119
FQ230964.1 - 7 - 8 670 119

View Full Product Details