Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx2
Entrez Gene ID
303398
Gene Name
T-box 2
Gene Aliases
-
Chromosome Location
Chr.10: 73278999 - 73289940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107033.1 NP_001100503.1 4 - 5 634 118

View Full Product Details