Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spag7
Entrez Gene ID
303260
Gene Name
sperm associated antigen 7
Gene Aliases
-
Chromosome Location
Chr.10: 57284989 - 57291146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107016.1 NP_001100486.1 5 - 6 552 89
GenBank mRNA FQ233279.1 - 5 - 6 424 89

View Full Product Details