The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108295.1 | NP_001101765.1 | 8 - 9 | 1082 | 53 |
GenBank mRNA | BC168194.1 | - | 6 - 7 | 790 | 53 |