Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snap47
Entrez Gene ID
303183
Gene Name
synaptosomal-associated protein, 47
Gene Aliases
RGD735194, SNAP-47
Chromosome Location
Chr.10: 45757196 - 45798752 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199389.1 NP_955421.1 4 - 5 1244 109
XM_006246488.2 XP_006246550.1 4 - 5 1278 109
GenBank mRNA BC062056.1 - 4 - 5 1244 109

View Full Product Details