Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cisd3
Entrez Gene ID
287661
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
Mel13, RGD1559720
Chromosome Location
Chr.10: 85628596 - 85631656 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105835.1 NP_001099305.1 3 - 4 536 72

View Full Product Details