Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thoc6
Entrez Gene ID
79227
Gene Name
THO complex 6 homolog (Drosophila)
Gene Aliases
Pdrp
Chromosome Location
Chr.10: 12989135 - 12994495 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024384.2 NP_077360.2 7 - 8 727 73
GenBank mRNA BC079149.1 - 7 - 8 727 73

View Full Product Details