The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013119.1 | NP_001013137.1 | 3 - 4 | 560 | 106 |
GenBank mRNA | AY553631.1 | - | 3 - 4 | 560 | 106 |
AY553632.1 | - | 3 - 4 | 560 | 106 | |
DQ120481.1 | - | 3 - 4 | 560 | 106 | |
DQ120482.1 | - | 3 - 4 | 560 | 106 |