Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkia
Entrez Gene ID
114906
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor alpha
Gene Aliases
-
Chromosome Location
Chr.2: 96593185 - 96668222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232153.2 XP_006232215.1 1 - 2 545 83

View Full Product Details