The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047885.1 | NP_001041350.1 | 6 - 7 | 995 | 106 |
XM_006232280.2 | XP_006232342.1 | 7 - 8 | 1182 | 106 | |
XM_006232281.2 | XP_006232343.1 | 7 - 8 | 1187 | 106 | |
GenBank mRNA | BC168644.1 | - | 5 - 6 | 2188 | 106 |