Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem86a
Entrez Gene ID
308602
Gene Name
transmembrane protein 86A
Gene Aliases
RGD1305749
Chromosome Location
Chr.1: 103173032 - 103177382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135016.1 NP_001128488.1 1 - 2 118 88
GenBank mRNA BE100466.1 - 2 - 3 88

View Full Product Details