Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rergl
Entrez Gene ID
500356
Gene Name
RERG/RAS-like
Gene Aliases
RGD1560090
Chromosome Location
Chr.4: 173630215 - 173640755 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191979.1 NP_001178908.1 4 - 5 377 114
XM_008763418.1 XP_008761640.1 3 - 4 373 114

View Full Product Details