Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec1b
Entrez Gene ID
500336
Gene Name
C-type lectin domain family 1, member B
Gene Aliases
RGD1563517
Chromosome Location
Chr.4: 163160680 - 163170470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191978.1 NP_001178907.1 3 - 4 437 117
XM_006237088.2 XP_006237150.1 3 - 4 2013 117
XM_006237089.2 XP_006237151.1 2 - 3 758 117
XM_006237090.2 XP_006237152.1 2 - 3 1915 117
GenBank mRNA FQ227184.1 - 3 - 4 506 117
FQ227324.1 - 3 - 4 506 117
GU357482.1 - 3 - 4 287 117

View Full Product Details