Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nanog
Entrez Gene ID
414065
Gene Name
Nanog homeobox
Gene Aliases
-
Chromosome Location
Chr.4: 155531906 - 155539268 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100781.1 NP_001094251.1 3 - 4 705 96

View Full Product Details