Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Necap1
Entrez Gene ID
312694
Gene Name
NECAP endocytosis associated 1
Gene Aliases
RGD1306053
Chromosome Location
Chr.4: 155709712 - 155724801 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029919.2 NP_001025090.1 2 - 3 216 102
XM_006237305.1 XP_006237367.1 3 - 4 277 102
GenBank mRNA BC097496.1 - 2 - 3 216 102

View Full Product Details