The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003750790.3 | XP_003750838.1 | 9 - 10 | 991 | 144 |
XM_006246064.2 | XP_006246126.1 | 8 - 9 | 1072 | 144 | |
XM_006246065.1 | XP_006246127.1 | 5 - 6 | 682 | 144 | |
XM_008767617.1 | XP_008765839.1 | 6 - 7 | 1080 | 144 |