The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004274.2 | NP_001004274.1 | 3 - 4 | 873 | 57 |
GenBank mRNA | AY686592.1 | - | 3 - 4 | 637 | 57 |
BC098750.1 | - | 3 - 4 | 873 | 57 |