The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271207.1 | NP_001258136.1 | 2 - 3 | 5804 | 65 |
XM_008767818.1 | XP_008766040.1 | 1 - 2 | 5518 | 65 | |
XM_008767819.1 | XP_008766041.1 | 2 - 3 | 6448 | 65 | |
GenBank mRNA | AY724505.1 | - | 1 - 2 | 644 | 65 |