Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rundc3a
Entrez Gene ID
303569
Gene Name
RUN domain containing 3A
Gene Aliases
GCRP, Rap2ip
Chromosome Location
Chr.10: 90342029 - 90351172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198758.2 NP_942053.2 3 - 4 612 67
XM_006247335.2 XP_006247397.1 3 - 4 634 67
XM_006247337.2 XP_006247399.1 3 - 4 634 67
XM_006247339.2 XP_006247401.1 3 - 4 634 67
XM_006247340.2 XP_006247402.1 3 - 4 619 67
XM_008768056.1 XP_008766278.1 3 - 4 622 67
XM_008768058.1 XP_008766280.1 3 - 4 619 67
XM_008768059.1 XP_008766281.1 3 - 4 634 67
GenBank mRNA BC127472.1 - 2 - 3 196 67
CB797318.1 - 2 - 3 293 67

View Full Product Details