Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rundc3a
Entrez Gene ID
303569
Gene Name
RUN domain containing 3A
Gene Aliases
GCRP, Rap2ip
Chromosome Location
Chr.10: 90342029 - 90351172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198758.2 NP_942053.2 5 - 6 783 64
XM_006247335.2 XP_006247397.1 5 - 6 805 64
XM_006247337.2 XP_006247399.1 5 - 6 805 64
XM_006247339.2 XP_006247401.1 5 - 6 805 64
XM_006247340.2 XP_006247402.1 5 - 6 790 64
XM_008768056.1 XP_008766278.1 5 - 6 793 64
XM_008768058.1 XP_008766280.1 5 - 6 790 64
XM_008768059.1 XP_008766281.1 5 - 6 805 64
GenBank mRNA BC127472.1 - 4 - 5 367 64
BC158572.1 - 2 - 3 180 64

View Full Product Details