Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eme2
Entrez Gene ID
302982
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
RGD1304924
Chromosome Location
Chr.10: 14253552 - 14256993 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750789.3 XP_003750837.1 3 - 4 522 106
XM_008767602.1 XP_008765824.1 3 - 4 522 106
XM_008767603.1 XP_008765825.1 3 - 4 525 106
XM_008767604.1 XP_008765826.1 3 - 4 525 106
XM_008767605.1 XP_008765827.1 3 - 4 488 106
GenBank mRNA BC158841.1 - 3 - 4 468 106

View Full Product Details