Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1e
Entrez Gene ID
360593
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1E
Gene Aliases
-
Chromosome Location
Chr.10: 74548270 - 74679858 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198773.1 NP_942068.1 3 - 4 894 112
GenBank mRNA AB081729.1 - 3 - 4 894 112

View Full Product Details