Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx11
Entrez Gene ID
303493
Gene Name
sorting nexin 11
Gene Aliases
-
Chromosome Location
Chr.10: 84628420 - 84639126 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012012.2 NP_001012012.2 4 - 5 292 77
NM_001199169.1 NP_001186098.1 3 - 4 232 77
XM_006247195.2 XP_006247257.1 5 - 6 685 77
XM_006247196.2 XP_006247258.1 4 - 5 462 77
XM_006247198.2 XP_006247260.1 3 - 4 402 77
XM_008768047.1 XP_008766269.1 5 - 6 1036 77
XM_008768048.1 XP_008766270.1 5 - 6 1186 77
GenBank mRNA BC086543.1 - 4 - 5 292 77
DN932564.1 - - 227 77
FQ212083.1 - 4 - 5 263 77

View Full Product Details