The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044224.1 | NP_001037689.1 | 23 - 24 | 3546 | 91 |
GenBank mRNA | AB231661.1 | - | 23 - 24 | 3546 | 91 |
AF111111.1 | - | 11 - 12 | 1118 | 91 | |
BC166408.1 | - | 23 - 24 | 3536 | 91 |