The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181377.3 | NP_852042.1 | 1 - 2 | 322 | 119 |
GenBank mRNA | BC129084.1 | - | 1 - 2 | 322 | 119 |