The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013859.1 | NP_001013881.1 | 8 - 9 | 1065 | 64 |
GenBank mRNA | BC083572.1 | - | 8 - 9 | 1065 | 64 |
FQ228522.1 | - | 6 - 7 | 1974 | 64 |