Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss34
Entrez Gene ID
287140
Gene Name
protease, serine, 34
Gene Aliases
-
Chromosome Location
Chr.10: 14656812 - 14658569 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105772.1 NP_001099242.1 1 - 2 162 79

View Full Product Details