Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa4
Entrez Gene ID
266759
Gene Name
heat shock protein 4
Gene Aliases
Hsp110, Hsp70, irp94
Chromosome Location
Chr.10: 38601624 - 38642441 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153629.1 NP_705893.1 5 - 6 755 87
XM_006246232.1 XP_006246294.1 5 - 6 799 87
GenBank mRNA AF077354.1 - 5 - 6 755 87
FQ222433.1 - 6 - 7 963 87

View Full Product Details