Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt14
Entrez Gene ID
287701
Gene Name
keratin 14, type I
Gene Aliases
Ka14, Krt1-14
Chromosome Location
Chr.10: 88118175 - 88122233 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008751.1 NP_001008751.1 3 - 4 787 60

View Full Product Details