Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc63
Entrez Gene ID
364427
Gene Name
leucine rich repeat containing 63
Gene Aliases
-
Chromosome Location
Chr.15: 57133036 - 57170467 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024803.1 NP_001019974.1 8 - 9 1549 150
XM_006252325.2 XP_006252387.1 8 - 9 1609 150
XM_006252326.2 XP_006252388.1 8 - 9 1643 150
XM_006252327.2 XP_006252389.1 8 - 9 1592 150
GenBank mRNA BC083605.1 - 8 - 9 1609 150
BC097406.1 - 8 - 9 1549 150

View Full Product Details