The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106051.3 | NP_001099521.1 | 4 - 5 | 315 | 71 |
NM_001270865.1 | NP_001257794.1 | 4 - 5 | 397 | 71 | |
NM_001270866.1 | NP_001257795.1 | 4 - 5 | 315 | 71 | |
GenBank mRNA | BC158790.1 | - | 4 - 5 | 306 | 71 |
FM108159.1 | - | 4 - 5 | 338 | 71 | |
FQ213565.1 | - | 4 - 5 | 315 | 71 | |
FQ218750.1 | - | 4 - 5 | 288 | 71 | |
FQ225558.1 | - | 1 - 2 | 75 | 71 | |
FQ229329.1 | - | 4 - 5 | 289 | 71 | |
FQ234562.1 | - | 1 - 2 | 75 | 71 |