Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdga2
Entrez Gene ID
314180
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
Mamdc1
Chromosome Location
Chr.6: 88446720 - 89292027 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199269.1 NP_954890.1 6 - 7 1162 141
XM_008764688.1 XP_008762910.1 5 - 6 1951 141
GenBank mRNA AY371924.1 - 6 - 7 1162 141

View Full Product Details