The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013097.2 | NP_001013115.1 | 2 - 3 | 309 | 87 |
NM_001286938.1 | NP_001273867.1 | 2 - 3 | 309 | 87 | |
GenBank mRNA | BC091209.1 | - | 2 - 3 | 284 | 87 |
CB585145.1 | - | 2 - 3 | 309 | 87 |