Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agmo
Entrez Gene ID
362732
Gene Name
alkylglycerol monooxygenase
Gene Aliases
RGD1312038, Tmem195
Chromosome Location
Chr.6: 56846859 - 57193918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135899.1 NP_001129371.1 2 - 3 388 66
GenBank mRNA BC127545.1 - 2 - 3 388 66
FQ211046.1 - 2 - 3 444 66
FQ219104.1 - 2 - 3 432 66

View Full Product Details