Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agmo
Entrez Gene ID
362732
Gene Name
alkylglycerol monooxygenase
Gene Aliases
RGD1312038, Tmem195
Chromosome Location
Chr.6: 56846859 - 57193918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135899.1 NP_001129371.1 12 - 13 1393 93
GenBank mRNA BC127545.1 - 12 - 13 1393 93
FQ211046.1 - 12 - 13 1449 93

View Full Product Details