The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108705.2 | NP_001102175.1 | 4 - 5 | 445 | 82 |
XM_006239970.2 | XP_006240032.1 | 4 - 5 | 877 | 82 | |
GenBank mRNA | BC158742.1 | - | 4 - 5 | 445 | 82 |