The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007613.1 | NP_001007614.1 | 11 - 12 | 1382 | 105 |
XM_008768312.1 | XP_008766534.1 | 10 - 11 | 1371 | 105 | |
XM_008768313.1 | XP_008766535.1 | 10 - 11 | 1294 | 105 | |
GenBank mRNA | BC079036.1 | - | 11 - 12 | 1382 | 105 |
FJ168767.1 | - | 10 - 11 | 1213 | 105 |