Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdpk1
Entrez Gene ID
81745
Gene Name
3-phosphoinositide dependent protein kinase-1
Gene Aliases
-
Chromosome Location
Chr.10: 13446373 - 13525248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031081.1 NP_112343.1 13 - 14 1621 76
XM_006246036.2 XP_006246098.1 13 - 14 3473 76
GenBank mRNA Y15748.1 - 13 - 14 1621 76

View Full Product Details