Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdpk1
Entrez Gene ID
81745
Gene Name
3-phosphoinositide dependent protein kinase-1
Gene Aliases
-
Chromosome Location
Chr.10: 13446373 - 13525248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031081.1 NP_112343.1 4 - 5 529 105
XM_006246036.2 XP_006246098.1 4 - 5 2381 105
XM_006246038.2 XP_006246100.1 4 - 5 591 105
GenBank mRNA Y15748.1 - 4 - 5 529 105

View Full Product Details