Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdpk1
Entrez Gene ID
81745
Gene Name
3-phosphoinositide dependent protein kinase-1
Gene Aliases
-
Chromosome Location
Chr.10: 13446373 - 13525248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031081.1 NP_112343.1 8 - 9 923 122
XM_006246036.2 XP_006246098.1 8 - 9 2775 122
XM_006246038.2 XP_006246100.1 8 - 9 985 122
GenBank mRNA Y15748.1 - 8 - 9 923 122

View Full Product Details