Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prr11
Entrez Gene ID
360591
Gene Name
proline rich 11
Gene Aliases
RGD1561541
Chromosome Location
Chr.10: 74393683 - 74434978 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108287.1 NP_001101757.1 4 - 5 465 69
XM_008768073.1 XP_008766295.1 4 - 5 622 69

View Full Product Details