The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108287.1 | NP_001101757.1 | 9 - 10 | 1095 | 63 |
XM_008768073.1 | XP_008766295.1 | 9 - 10 | 1252 | 63 |