Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sirt7
Entrez Gene ID
303745
Gene Name
sirtuin 7
Gene Aliases
-
Chromosome Location
Chr.10: 109796046 - 109802791 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107073.1 NP_001100543.1 6 - 7 607 73
XM_006247889.2 XP_006247951.1 6 - 7 656 73
GenBank mRNA BC167031.1 - 6 - 7 589 73

View Full Product Details