The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017225.2 | NP_058921.1 | 4 - 5 | 509 | 74 |
XM_006247124.2 | XP_006247186.1 | 4 - 5 | 495 | 74 | |
XM_008767982.1 | XP_008766204.1 | 6 - 7 | 716 | 74 | |
XM_008767983.1 | XP_008766205.1 | 6 - 7 | 670 | 74 | |
GenBank mRNA | AF040261.1 | - | 4 - 5 | 453 | 74 |
BC078854.1 | - | 4 - 5 | 514 | 74 |