Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nup85
Entrez Gene ID
287830
Gene Name
nucleoporin 85
Gene Aliases
Pcnt1
Chromosome Location
Chr.10: 104118999 - 104137258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025401.1 NP_001020572.1 2 - 3 151 63
GenBank mRNA BC098031.1 - 2 - 3 151 63

View Full Product Details