Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clk4
Entrez Gene ID
287269
Gene Name
CDC-like kinase 4
Gene Aliases
-
Chromosome Location
Chr.10: 36715565 - 36733476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013041.1 NP_001013059.1 5 - 6 596 113
XM_006246234.2 XP_006246296.1 6 - 7 806 113
XM_006246236.1 XP_006246298.1 9 - 10 1009 113
XM_006246237.1 XP_006246299.1 7 - 8 711 113
XM_006246238.2 XP_006246300.1 6 - 7 2226 113
XM_008767670.1 XP_008765892.1 7 - 8 1680 113
XM_008767671.1 XP_008765893.1 6 - 7 1602 113
XM_008767672.1 XP_008765894.1 6 - 7 645 113
XM_008767673.1 XP_008765895.1 5 - 6 2298 113
XM_008767674.1 XP_008765896.1 4 - 5 2085 113
XM_008767675.1 XP_008765897.1 7 - 8 872 113
XM_008767676.1 XP_008765898.1 5 - 6 597 113
GenBank mRNA BC079006.1 - 5 - 6 596 113
FQ223275.1 - 8 - 9 2656 113

View Full Product Details